Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF131 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111722
Description
Immunogen sequence: GRKKLYECQV CNSVFNSWDQ FKDHLVIHTG DKPNHCTLCD LWFMQGNELR RHLSDAHNIS ERLVTEEVLS VETRVQTEPV TSMTIIEQVG KVHVLPLLQV QVDSAQVTVE QVHPDLLQD.
May be involved in transcriptional regulation. Plays a role during development and organogenesis as well as in the function of the adult central nervous system.
Specifications
ZNF131 | |
Polyclonal | |
Unconjugated | |
ZNF131 | |
2610109I01Rik; AW558871; pHZ-10; ZBTB35; Zfp131; zinc finger and BTB domain containing 35; zinc finger protein 131; zinc finger protein 131 (clone pHZ-10); Zinc finger protein 131-like protein; ZNF131 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7690 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.20 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P52739 | |
ZNF131 | |
Recombinant Protein corresonding to Human ZNF131. Recombinant protein control fragment (Product #RP-108011). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction