Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF165 Antibody (OTI4C1), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N274932A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274932A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274932A700 Supplier Novus Biologicals Supplier No. NBP274932AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF165 Monoclonal specifically detects ZNF165 in Human samples. It is validated for Western Blot.

Specifications

Antigen ZNF165
Applications Western Blot
Classification Monoclonal
Clone OTI4C1
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cancer/testis antigen 53, CT53Zinc finger and SCAN domain-containing protein 7, zinc finger protein 165, ZSCAN7LD65ZPF165
Gene Symbols ZNF165
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 165-485 of human ZNF165 (NP_003438) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7718.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.