Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF197 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB034073 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB034073 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB034073 Supplier Novus Biologicals Supplier No. NBP2864490.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF197 Polyclonal specifically detects ZNF197 in Human samples. It is validated for Western Blot.

Specifications

Antigen ZNF197
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias D3S1363E, P18, VHL-associated KRAB-A domain-containing protein, zinc finger protein 166, zinc finger protein 197, zinc finger protein 20, Zinc finger protein with KRAB and SCAN domains 9, ZKSCAN9VHLaK, ZNF166, ZNF20
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N-terminal region of human ZNF197
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10168
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.