Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF205 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB181304 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB181304 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB181304 Supplier Novus Biologicals Supplier No. NBP306486F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNF205 Polyclonal antibody specifically detects ZNF205 in Human samples. It is validated for Immunofluorescence

Specifications

Antigen ZNF205
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias zinc finger protein 205, Zinc finger protein 210, ZNF210Zfp13
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ZNF205 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7755
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.