Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF217 Antibody (OTI3G5), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N274933J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274933J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274933J646 Supplier Novus Biologicals Supplier No. NBP274933JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF217 Monoclonal specifically detects ZNF217 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen ZNF217
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3G5
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp781O2147, KIAA0335, MGC125707, MGC125710, zinc finger protein 518, zinc finger protein 518A, ZNF518
Gene Symbols ZNF217
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 730-1048 of human ZNF217 (NP_006517) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7764.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.