Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF230 Antibody (OTI1E2), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB274934A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB274934A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB274934A48 Supplier Novus Biologicals Supplier No. NBP274934AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF230 Monoclonal specifically detects ZNF230 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen ZNF230
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI1E2
Conjugate Alexa Fluor 488
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ12515, FLJ14855, zinc finger protein 502
Gene Symbols ZNF230
Host Species Mouse
Immunogen Full length recombinant protein of human ZNF230 (NP_006291) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 7773.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.