Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF230 Antibody (OTI1E2), Novus Biologicals™
SDP

Catalog No. NBP246425 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP246425 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP246425 Supplier Novus Biologicals Supplier No. NBP246425
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Immunohistochemistry

ZNF230 Monoclonal specifically detects ZNF230 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ZNF230
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI1E2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:150
Formulation PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
Gene Alias FLJ12515, FLJ14855, zinc finger protein 502
Gene Symbols ZNF230
Host Species Mouse
Immunogen Full length human recombinant protein of human ZNF230 (NP_006291) produced in E.coli.
Molecular Weight of Antigen 54.4 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 7773
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.