Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF259 Antibody (LG1), HRP, Novus Biologicals™
SDP

Catalog No. NBP222448H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222448H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222448H Supplier Novus Biologicals Supplier No. NBP222448H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF259 Monoclonal specifically detects ZNF259 in Human samples. It is validated for Western Blot.

Specifications

Antigen ZNF259
Applications Western Blot
Classification Monoclonal
Clone LG1
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias ERP1, EZFIT-related protein 1, KIAA1396MGC150657, MGC150658, Z1971, zinc finger protein 471
Gene Symbols ZNF259
Host Species Mouse
Immunogen murine ZPR1 protein
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8882
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.