Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF266 Antibody, Novus Biologicals™
SDP

Catalog No. NB396167 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396167 25 μL
NBP247567 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396167 Supplier Novus Biologicals Supplier No. NBP24756725UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF266 Polyclonal specifically detects ZNF266 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ZNF266
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias KOX5BRC1744, Zinc finger protein 13Zinc finger protein KOX5, zinc finger protein 45, zinc finger protein 45 (a Kruppel-associated box (KRAB) domain polypeptide), ZNF13
Gene Symbols ZNF266
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: NLATVGYQLFKPSLISWLEQEESRTVQRGDFQASEWKVQLKTKELALQQDVLGEPTSSGIQMIGSHNGGEVSDVKQCGDVSSEHSCLKTHV
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10781
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.