Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF280D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541154
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541154 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541154 Supplier Thermo Scientific Supplier No. PA541154
Only null left
Add to Cart
Add to Cart

ZNF280D Polyclonal Antibody

ZNF280D Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF280D
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF280D
Gene Accession No. Q6N043
Gene Alias FLJ20086; KIAA1584; SUHW4; Suppressor of hairy wing homolog 4; zinc finger protein 280D; Zinc finger protein 634; ZNF280D; ZNF634
Gene Symbols ZNF280D
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZNF280D.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54816
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.