Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF286 Antibody (OTI7D2), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP274935FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274935FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274935FR Supplier Novus Biologicals Supplier No. NBP274935FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF286 Monoclonal specifically detects ZNF286 in Human samples. It is validated for Western Blot.

Specifications

Antigen ZNF286
Applications Western Blot
Classification Monoclonal
Clone OTI7D2
Conjugate DyLight 680
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias MGC149627, zinc finger protein 286, zinc finger protein 286A
Gene Symbols ZNF286A
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-252 of human ZNF286 (NP_065703) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 57335.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.