Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF334 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$480.74

Specifications

Antigen ZNF334
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP179707
SDP
View Documents
Novus Biologicals
NBP179707
100 μL
Each of 1 for $480.74
Only null left
Add to Cart
 
Description

Description

ZNF334 Polyclonal specifically detects ZNF334 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZNF334
Polyclonal
Rabbit
NP_060572
55713
Synthetic peptide directed towards the N terminal of human ZNF334The immunogen for this antibody is ZNF334. Peptide sequence KMKKFQIPVSFQDLTVNFTQEEWQQLDPAQRLLYRDVMLENYSNLVSVGY.
Primary
Western Blot
Unconjugated
RUO
DKFZp586G1122, Hzf, HZFzinc finger protein 385, RZFHematopoietic zinc finger protein, ZFP385, zinc finger protein 385A, ZNF385Retinal zinc finger protein
ZNF334
IgG
80 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.