Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF34 Antibody (OTI3G2), HRP, Novus Biologicals™
SDP

Catalog No. NBP274937H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274937H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274937H Supplier Novus Biologicals Supplier No. NBP274937H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF34 Monoclonal specifically detects ZNF34 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen ZNF34
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3G2
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias FLJ3472, KOX21zinc finger protein 37a (KOX 21), zinc finger protein 37A, Zinc finger protein KOX21, ZNF37
Gene Symbols ZNF34
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 117-405 of human ZNF34 (NP_085057) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 80778.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.