Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF35 Antibody (OTI4D5), Biotin, Novus Biologicals™
SDP

Catalog No. NBP274938B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274938B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274938B Supplier Novus Biologicals Supplier No. NBP274938B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF35 Monoclonal specifically detects ZNF35 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen ZNF35
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4D5
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias BRF2, Butyrate response factor 2, ERF2ERF-2, Protein TIS11D, RNF162Czinc finger protein 36, C3H1 type-like 2, TIS11DEGF-response factor 2, zinc finger protein 36, C3H type-like 1, zinc finger protein 36, C3H type-like 2, zinc finger protein, C3H type, 36-like 2
Gene Symbols ZNF35
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-258 of human ZNF35 (NP_003411) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7584.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.