Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF350 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176344 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176344 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176344 Supplier Novus Biologicals Supplier No. NBP176344
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF350 Polyclonal specifically detects ZNF350 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen ZNF350
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_067645
Gene Alias Berg36, BRF1zinc finger protein 36, C3H1 type-like 1, Butyrate response factor 1, cMG1, ERF1ERF-1, Protein TIS11B, RNF162BEGF-response factor 1, TIS11Bearly response factor Berg36, zinc finger protein 36, C3H type-like 1, zinc finger protein, C3H type, 36-like 1
Gene Symbols ZNF350
Host Species Rabbit
Immunogen Antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of the human ZBRK1. The immunogen is located within amino acids 410 - 460 of ZBRK1. Amino Acid Squence: KRIHTREKQEAKKVENPPAE
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer
Primary or Secondary Primary
Gene ID (Entrez) 59348
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.