Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF354A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | ZNF354A |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-9191
![]()
|
Proteintech Group Inc
140721AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
ZNF354A gene ontology annotations related to this gene include regulation of transcription by RNA polymerase II; sensory perception of sound.Specifications
| ZNF354A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O60765, Q02975, Q61751 | |
| 21408, 24522, 6940 | |
| ZNF354A Fusion Protein Ag5142 | |
| Primary | |
| -20°C | |
| ZNF354A |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| EZNF, HKL1, KID 1, KID1, TCF 17, TCF17, Transcription factor 17, zinc finger protein 354A, Zinc finger protein eZNF, ZNF354A | |
| ZNF354A | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title