Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF382 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$499.50

Specifications

Antigen ZNF382
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP180380
SDP
View Documents
Novus Biologicals
NBP180380
100 μL
Each for $499.50
Only null left
Add to Cart
 
NBP18038020
SDP
View Documents
Novus Biologicals
NBP18038020UL
20 μL N/A N/A N/A
Description

Description

ZNF382 Polyclonal specifically detects ZNF382 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZNF382
Polyclonal
Purified
RUO
zinc finger protein 382
ZNF382
IgG
Protein A purified
Western Blot
Unconjugated
Rabbit
NP_116214
84911
Synthetic peptide directed towards the N terminal of human ZNF382. Peptide sequence MPLQGSVSFKDVTVDFTQEEWQQLDPAQKALYRDVMLENYCHFVSVGFHM.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.