Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF385B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | ZNF385B |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ZNF385B Polyclonal specifically detects ZNF385B in Human samples. It is validated for Western Blot.Specifications
| ZNF385B | |
| Polyclonal | |
| Rabbit | |
| NP_001106869 | |
| 151126 | |
| Synthetic peptide directed towards the C terminal of human ZNF385BThe immunogen for this antibody is ZNF385B. Peptide sequence HVNSEIQLKQHISSRRHKDRVAGKPLKPKYSPYNKLQRSPSILAAKLAFQ. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| C2H2-like zinc finger protein rearranged in thyroid adenomas, DKFZp686L0787, KRAB zinc finger protein, zinc finger protein 331, Zinc finger protein 463, ZNF463RITAZNF361Zinc finger protein 361 | |
| ZNF385B | |
| IgG | |
| 41 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title