Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF385B Antibody (OTI1H10), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N274948A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274948A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274948A405 Supplier Novus Biologicals Supplier No. NBP274948AF405

Mouse Monoclonal Antibody

ZNF385B Monoclonal specifically detects ZNF385B in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen ZNF385B
Applications Western Blot
Classification Monoclonal
Clone OTI1H10
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C2H2-like zinc finger protein rearranged in thyroid adenomas, DKFZp686L0787, KRAB zinc finger protein, zinc finger protein 331, Zinc finger protein 463, ZNF463RITAZNF361Zinc finger protein 361
Gene Symbols ZNF385B
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 149-399 of human ZNF385B (NP_689733) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 151126.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.