Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF385B Monoclonal Antibody (OTI2F3), TrueMAB™, OriGene
SDP

Catalog No. 501679386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-9386 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-9386 Supplier OriGene Supplier No. TA802748
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF385B contains 4 U1-type zinc fingers. ZNF385B is expressed in brain with a potential role in RNA maturation, stability, or transcription regulation. There are 5 known isoforms.

Specifications

Antigen ZNF385B
Applications Western Blot
Classification Monoclonal
Clone OTI2F3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ZNF385B
Gene Accession No. Q569K4
Gene Alias ZNF533
Gene Symbols ZNF385B
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 149-399 of human ZNF385B produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151126
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.