Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF385D Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB327527 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB327527 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB327527 Supplier Novus Biologicals Supplier No. NBP305996APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF385D Polyclonal antibody specifically detects ZNF385D in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen ZNF385D
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias FLJ12586, zinc finger protein 329
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ZNF385D fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79750
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.