Learn More
Description
Specifications
Specifications
| Antigen | ZNF385D |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 549 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FLJ12586, zinc finger protein 329 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human ZNF385D fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
