Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF394 Antibody (OTI1G9), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB274949A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB274949A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB274949A48 Supplier Novus Biologicals Supplier No. NBP274949AF488

Mouse Monoclonal Antibody

ZNF394 Monoclonal specifically detects ZNF394 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen ZNF394
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1G9
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias dJ874C20.2, FLJ23407, zinc finger protein 310 pseudogene, zinc finger protein 323, ZNF20-Lp, ZNF310P
Gene Symbols ZNF394
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-296 of human ZNF394 (NP_115540) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 84124.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.