Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF397 Antibody (OTI1F3), FITC, Novus Biologicals™
SDP

Catalog No. NBP274950F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274950F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274950F Supplier Novus Biologicals Supplier No. NBP274950F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF397 Monoclonal specifically detects ZNF397 in Human, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ZNF397
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1F3
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias KIAA1588, KRAB-containing zinc finger protein 317, zinc finger protein 317
Gene Symbols ZNF397
Host Species Mouse
Immunogen Full length recombinant protein of human ZNF397 (NP_115723) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 84307.0
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.