Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF407 Antibody, Novus Biologicals™
SDP

Catalog No. NBP234850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234850 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234850 Supplier Novus Biologicals Supplier No. NBP234850
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF407 Polyclonal specifically detects ZNF407 in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen ZNF407
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2 - 10 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_060227.2
Gene Alias FLJ13839, FLJ20307, KIAA1703, zinc finger protein 407
Gene Symbols ZNF407
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 2000 to 2050 of human Zinc Finger Protein 407 using the numbering given in entry NP_060227.2 (GeneID 55628).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55628
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla and Chimpanzee.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.