Learn More
Description
Specifications
Specifications
| Antigen | ZNF416 |
| Applications | Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | FLJ12859, FLJ14378, GC-box-binding zinc finger protein 1, GZP1, Transcription factor ZBP-99, ZBP99, ZBP-99, Zinc finger DNA-binding protein 99, zinc finger protein 281, ZNP-99, ZNP-99 transcription factor |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 495-594 of human ZNF416 (NP_060349.1). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
