Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF423 Antibody, Novus Biologicals™
SDP

Catalog No. NBP276351 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP276351 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP276351 Supplier Novus Biologicals Supplier No. NBP276351
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ZNF423 Polyclonal specifically detects ZNF423 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.

Specifications

Antigen ZNF423
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 10 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Peptide ELISA Detection limit 1:2000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias FLJ20086, KIAA1584, MGC21637, MGC61687, SUHW4, Suppressor of hairy wing homolog 4, zinc finger protein 280D, Zinc finger protein 634, ZNF634suppressor of hairy wing homolog 4 (Drosophila)
Gene Symbols ZNF423
Host Species Goat
Immunogen Peptide with sequence EPECDQKTSRALEDR corresponding to N-Terminus (near) according to NP_055884.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Primary or Secondary Primary
Gene ID (Entrez) 23090.0
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.