Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF432 Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | ZNF432 |
---|---|
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111346
|
Invitrogen™
PA5111346 |
100 μL |
Each for $589.50
|
|
|||||
PIPA5111602
|
Invitrogen™
PA5111602 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: SCEINNSTKF SGDGKSFLHG NYEELYSAAK FSVSTKANST KSQVSKHQRT HEI.
ZNF432 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 16 C2H2-type zinc fingers and 1 KRAB domain. ZNF432 may be involved in transcriptional regulation.Specifications
ZNF432 | |
Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94892 | |
9668 | |
Recombinant Protein corresonding to Human ZNF432. Recombinant protein control fragment (Product #RP-100010). | |
Antibody |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
ZNF432 | |
KIAA0798; Zinc finger protein 432; ZNF432 | |
ZNF432 | |
Primary | |
Antigen affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title