Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF433 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179369
Description
ZNF433 Polyclonal specifically detects ZNF433 in Human samples. It is validated for Western Blot.Specifications
ZNF433 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FLJ38685, FLJ77809, MGC45417, zfp-276, ZFP276MGC126480, zinc finger protein 276, zinc finger protein 276 homolog, zinc finger protein 276 homolog (mouse), Zinc finger protein 477, ZNF477MGC111410 | |
Rabbit | |
Affinity purified | |
RUO | |
163059 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_001073880 | |
ZNF433 | |
Synthetic peptide directed towards the N terminal of human ZNF433The immunogen for this antibody is ZNF433. Peptide sequence GEVGSAHSSLNRHIRDDTGHKAYEYQEYGQKPYKCKYCKKPFNCLSSVQT. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%. | |
Human, Pig, Canine, Equine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction