Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF444 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB290953 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290953 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290953 Supplier Novus Biologicals Supplier No. NBP178190APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF444 Polyclonal antibody specifically detects ZNF444 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen ZNF444
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias HZF3, MGC126498, zinc finger protein 268, zinc finger protein 3, Zinc finger protein HZF3
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 150 and 200 of human Zinc Finger Protein 444 using the numbering given in entry NP_060807.2 (GeneID 55311).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55311
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.