Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF447 Antibody (OTI2H7), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP274951G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274951G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274951G Supplier Novus Biologicals Supplier No. NBP274951G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF447 Monoclonal specifically detects ZNF447 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ZNF447
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2H7
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp586B1122, FLJ12895, FLJ38919, FLJ44152, MGC4074, MGC8682, zinc finger and SCAN domain containing 18, zinc finger and SCAN domain-containing protein 18, Zinc finger protein 447MGC2427, ZNF447
Gene Symbols ZSCAN18
Host Species Mouse
Immunogen Full length recombinant protein of human ZNF447 (NP_076415) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 65982.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.