Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF468 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179437
Description
ZNF468 Polyclonal specifically detects ZNF468 in Human samples. It is validated for Western Blot.Specifications
ZNF468 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
ZABC1, zinc finger protein 217 | |
Rabbit | |
60 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Onchocerca volvulus: 100%; Canine: 100%; Rat: 100%; Florida lancelet: 100%; Trichoplax reptans: 100%; Bovine: 100%; Green puffer: 92%; Crab-eating macaque: 92%; Western clawed frog: 92%; Camponotus floridanus: 92%; Body louse: 92%; Chicken: 85%. | |
Human, Mouse, Rat, Bovine, Equine, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_001008801 | |
ZNF468 | |
Synthetic peptide directed towards the middle region of human ZNF468The immunogen for this antibody is ZNF468. Peptide sequence FIHKALHTGEKPYECEECDKVFSRKSHLERHKRIHTGEKPYKCKVCDEAF. | |
Affinity purified | |
RUO | |
90333 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction