Learn More
Description
Specifications
Specifications
| Antigen | ZNF483 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | BMZF-2zinc finger 2, bone marrow, BMZF2zinc finger protein 27 (KOX 22), Bone marrow zinc finger 2, KOX22Zinc finger protein 233, zinc finger protein 224, Zinc finger protein 255Zinc finger protein KOX22, Zinc finger protein 27, ZNF255zinc finger protein ZNF255, ZNF27ZNF233 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human ZNF483 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
