Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF486 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179462
Description
ZNF486 Polyclonal specifically detects ZNF486 in Human samples. It is validated for Western Blot.Specifications
ZNF486 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
KOX16MGC57283, Zfp612, zinc finger protein 23, zinc finger protein 23 (KOX 16), zinc finger protein 32, Zinc finger protein 359ZNF612kruppel-like zinc finger factor X31, Zinc finger protein 612, Zinc finger protein KOX16, ZNF359 | |
Rabbit | |
54 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Pig: 86%; Bovine: 86%; Dog: 85%; Horse: 85%; Rabbit: 85%; Zebrafish: 85%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_443084 | |
ZNF486 | |
Synthetic peptide directed towards the C terminal of human ZNF486The immunogen for this antibody is ZNF486. Peptide sequence HRRTHTGEKPYKCEECGKAYTTSSNLTEHKTTHTGEKPYKCKECGKAFNW. | |
Affinity purified | |
RUO | |
90649 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction