Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF487P Antibody, Novus Biologicals™
SDP

Catalog No. NBP193560 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP193560 0.1 mL
NB405229 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP193560 Supplier Novus Biologicals Supplier No. NBP193560
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF487P Polyclonal specifically detects ZNF487P in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ZNF487P
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias KRBO1, ZNF487, ZNF487P zinc finger protein 487, pseudogene
Gene Symbols ZNF487
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:EVVCKLEHGQVLWILEEESPSQSHLDCCIDDDLMEKRQENQDQHLQKVDFVNNKTLTMDRNGVLGKTFSLDTNPILSRKIRGNCDSSG
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 642819
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.