Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF500 Antibody (OTI2D2), HRP, Novus Biologicals™
SDP

Catalog No. NBP274953H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274953H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274953H Supplier Novus Biologicals Supplier No. NBP274953H

Mouse Monoclonal Antibody

ZNF500 Monoclonal specifically detects ZNF500 in Human samples. It is validated for Western Blot.

Specifications

Antigen ZNF500
Applications Western Blot
Classification Monoclonal
Clone OTI2D2
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias C2H2-171, RP58Zinc finger and BTB domain-containing protein 18, TAZ1, TAZ-1Zinc finger protein C2H2-171, Transcriptional repressor RP58, Translin-associated zinc finger protein 1, ZBTB1858 kDa repressor protein, zinc finger protein 238
Gene Symbols ZNF500
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 102-325 of human ZNF500 (NP_067678) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 26048.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.