Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF521 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB198454 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB198454 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB198454 Supplier Novus Biologicals Supplier No. NBP241174V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF521 Polyclonal antibody specifically detects ZNF521 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen ZNF521
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias EHZF, Evi3, LIP3, ZFP521, zinc finger protein 263, Zinc finger protein FPM315, Zinc finger protein with KRAB and SCAN domains 12
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human ZNF521. The immunogen is located within the first 50 amino acids of ZNF521. Amino Acid Squence: KKRPEDGEELEDEAVHS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 25925
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.