Learn More
Description
Specifically detects ZNF556 in Human samples, and it is validated for Immunocytochemistry, Immunofluorescence, Western Blotting
Specifications
Specifications
| Antigen | ZNF556 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 |
| Formulation | PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal |
| Gene Alias | FLJ11637, zinc finger protein 556 |
| Gene Symbols | ZNF556 |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the center region of human ZNF556. The exact sequence is proprietary. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
