Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF561 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18014820UL
Description
ZNF561 Polyclonal specifically detects ZNF561 in Human samples. It is validated for Western Blot.Specifications
ZNF561 | |
Polyclonal | |
Western Blot 1:1000 | |
NP_689502 | |
ZNF561 | |
Synthetic peptide directed towards the C terminal of human ZNF561. Peptide sequence KKPYQCKECGKAFTTSTSLIQHTRIHTGEKPYECVECGKTFITSSRRSKH. | |
Affinity Purified | |
RUO | |
93134 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS & 2% Sucrose. with No Preservative | |
MGC45408, zinc finger protein 561 | |
Rabbit | |
47 kDa | |
20 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction