Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF561 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | ZNF561 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18014820
![]() |
Novus Biologicals
NBP18014820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP180148
![]() |
Novus Biologicals
NBP180148 |
100 μL |
Each for $487.50
|
|
|||||
Description
ZNF561 Polyclonal specifically detects ZNF561 in Human samples. It is validated for Western Blot.Specifications
ZNF561 | |
Polyclonal | |
Rabbit | |
NP_689502 | |
93134 | |
Synthetic peptide directed towards the C terminal of human ZNF561. Peptide sequence KKPYQCKECGKAFTTSTSLIQHTRIHTGEKPYECVECGKTFITSSRRSKH. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
MGC45408, zinc finger protein 561 | |
ZNF561 | |
IgG | |
47 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title