Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF582 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB096236 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096236 20 μL
NB096235 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB096236 Supplier Novus Biologicals Supplier No. NBP30501220UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF582 Polyclonal specifically detects ZNF582 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen ZNF582
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias FLJ30927, zinc finger protein 582
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 40-180 of human ZNF582 (NP_653291.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147948
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.