Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF583 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
Antigen | ZNF583 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18016220
![]() |
Novus Biologicals
NBP18016220UL |
20 μL |
Each for $158.00
|
|
|||||
NBP180162
![]() |
Novus Biologicals
NBP180162 |
100 μL |
Each for $487.50
|
|
|||||
Description
ZNF583 Polyclonal specifically detects ZNF583 in Human samples. It is validated for Western Blot.Specifications
ZNF583 | |
Polyclonal | |
Rabbit | |
Human | |
FLJ31030, zinc finger protein 583 | |
ZNF583 | |
IgG | |
66 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_689691 | |
147949 | |
Synthetic peptide directed towards the N terminal of human ZNF583. Peptide sequence TRGPCPDWEYVFKNSEFSSKQETYEESSKVVTVGARHLSYSLDYPSLRED. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title