Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF607 Antibody, Novus Biologicals™
SDP

Catalog No. NBP1802520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1802520 20 μL
NBP180125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1802520 Supplier Novus Biologicals Supplier No. NBP18012520UL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

ZNF607 Polyclonal specifically detects ZNF607 in Human samples. It is validated for Western Blot.

Specifications

Antigen ZNF607
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. NP_116078
Gene Alias FLJ14802, MGC13071, zinc finger protein 607
Gene Symbols ZNF607
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZNF607. Peptide sequence KECGKGFTCRYQLTMHQRIYSGEKHYECKENGEAFSSGHQLTAPHTFESV.
Molecular Weight of Antigen 81 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84775
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.