Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF622 Antibody, Novus Biologicals™
SDP

Catalog No. NBP232102 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232102 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP232102 Supplier Novus Biologicals Supplier No. NBP232102
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF622 Polyclonal specifically detects ZNF622 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen ZNF622
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 mcg/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_219482.1
Gene Alias MGC2485, zinc finger protein 622, Zinc finger-like protein 9, ZPR9MGC17552
Gene Symbols ZNF622
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 125 to 175 of human Zinc Finger Protein 622 using the numbering given in entry NP_219482.1 (GeneID 90441).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90441
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.