Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF622 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB290095 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290095 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290095 Supplier Novus Biologicals Supplier No. NBP232087PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF622 Polyclonal antibody specifically detects ZNF622 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen ZNF622
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias MGC2485, zinc finger protein 622, Zinc finger-like protein 9, ZPR9MGC17552
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 25 to 75 of human Zinc Finger Protein 622 using the numbering given in entry NP_219482.1 (GeneID 90441).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90441
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.