Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF655 Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB362796 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362796 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362796 Supplier Novus Biologicals Supplier No. NBP178732IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF655 Polyclonal antibody specifically detects ZNF655 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen ZNF655
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias zinc finger protein 655
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1 and 50 of human Vav-1 Interacting Kruppel-like Protein using the numbering given in entry NP_612503.1 (GeneID 79027).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79027
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.