Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF664 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
Antigen | ZNF664 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179467
![]() |
Novus Biologicals
NBP179467 |
100 μL |
Each for $499.50
|
|
|||||
NBP17946720
![]() |
Novus Biologicals
NBP17946720UL |
20 μL | N/A | N/A | N/A | ||||
Description
ZNF664 Polyclonal specifically detects ZNF664 in Human samples. It is validated for Western Blot.Specifications
ZNF664 | |
Polyclonal | |
Rabbit | |
NP_689650 | |
144348 | |
Synthetic peptide directed towards the N terminal of human ZNF664The immunogen for this antibody is ZNF664. Peptide sequence FFHISELHIHWRDHTGEKVYKCDDCGKDFSTTTKLNRHKKIHTVEKPYKC. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
DKFZp761B128, MGC126579, ZFOC1zinc finger Organ of Corti 1, Zinc finger protein 176ZNF176, zinc finger protein 664, Zinc finger protein from organ of Corti, ZNF196 | |
ZNF664 | |
IgG | |
30 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title