Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF687 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB435978 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB435978 0.025 mg
NBP241128 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB435978 Supplier Novus Biologicals Supplier No. NBP2411280.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF687 Polyclonal specifically detects ZNF687 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ZNF687
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias KIAA1441DKFZp781I1719, zinc finger protein 687
Gene Symbols ZNF687
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human ZNF687. The immunogen is located within the first 50 amino acids of ZNF687. Amino Acid Squence: HSGPEENEGPGGPGKPE
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57592
Test Specificity ZNF687 antibody is human specific. At least three isoforms of ZNF687 are known to exist; this antibody will detect the the two largest isoforms.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.