Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF69 Antibody (OTI3C3), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP274954FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274954FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274954FR Supplier Novus Biologicals Supplier No. NBP274954FR

Mouse Monoclonal Antibody

ZNF69 Monoclonal specifically detects ZNF69 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen ZNF69
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3C3
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Cos5, hZNF3, MGC59928, zinc finger protein 69, zinc finger protein 69 (Cos5)
Gene Symbols ZNF69
Host Species Mouse
Immunogen Full length recombinant protein of human ZNF69 (NP_068734) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7620.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.