Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF690 Antibody (ZSCAN29/2610), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB011719 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB011719 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB011719 Supplier Novus Biologicals Supplier No. NBP279881AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF690 Monoclonal specifically detects ZNF690 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ZNF690
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ZSCAN29/2610
Conjugate Alexa Fluor 647
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.1.1.43, EC 2.7.10.1, FLJ35867, MGC129894, MGC129895, Zfp690, zinc finger and SCAN domain containing 29, zinc finger and SCAN domain-containing protein 29, Zinc finger protein 690KOX31-like zinc finger protein, ZNF690
Gene Symbols ZSCAN29
Host Species Mouse
Immunogen Recombinant full-length human ZNF690 protein (Uniprot: Q8IWY8)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.